Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
BluEcoli™ Urine Biplate, Hardy Diagnostics
Supplier: Hardy Diagnostics
J123
89501-514PK 59.94 USD InStock
89501-514
BluEcoli™ Urine Biplate, Hardy Diagnostics
Culture Dishes Petri Dishes
BluEcoli™ Urine Biplate (Blood Agar/Chromogenic E. coli), for detection of E. coli.
  • Primary plating medium when culturing urine specimens
  • Culture and isolation of urinary pathogens
  • Blue color detection of E. coli
  • Helpful in spotting mixed infections
A revolutionary agar bi-plate for screening urine specimens for E. coli, which causes about 85% of all urinary tract infections (UTI). Blood Agar (side 1) is recommended as a primary plating medium when culturing urine specimens.
BluEcoli™ (side 2) is a urine culture medium used for the culture and isolation of urinary pathogens and for the identification of E. coli which turns blue within 24 hours by using chromogenic substrates.
Inoculate both sides of the biplate with the urine specimen. If the infecting organism is E. coli, the colonies on the chromogenic side of the biplate will turn blue.
With the BluEcoli™ Urine Biplate, you can select a colony from the blood agar side of the biplate for susceptibility testing.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty