Would you like to log in again?
GLOVE NEO/LTX 28MIL S PK12/144
Supplier: PIP
Neoprene Unsupported Gloves^ Heavy 28 mil., blue neoprene over yellow latex, double dipped, flock lined, diamond grip, 12 inch length. Superior protection from a wide range of chemicals. Straight cuff. Provides chemical resistance properties of neoprene and the comfort of latex. Large.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
GLOVE NEO/LTX 28MIL S PK12/144 89501-660 52-3670/S Case of 144 Retrieving
Pack of 12 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty