Would you like to log in again?
Nalgene® Pharma-Grade Platinum-Cured Silicone Tubing, Thermo Scientific
Supplier: Thermo Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Nalgene®
8600-0190 8600-3025 8600-3024 8600-0140 8600-0260 8600-0070 8600-0100 8600-3016 8600-0310 8600-3017 8600-0050 8600-3018 8600-0120 8600-0060 8600-0080 8600-3173 8600-3015 8600-0040 8600-0200 8600-0020 8600-0030 8600-0130 8600-3182
89501-690CS 865.32 USD InStock
89501-690 89501-692 89501-670 89501-680 89501-676 89501-698 89501-686 89501-700 89501-678 89501-688 89501-666 89501-694 89501-672 89501-682 89501-674 89501-696 89501-684 89501-706 89501-702 89501-668 89501-704 89493-606 89493-604
Nalgene® Pharma-Grade Platinum-Cured Silicone Tubing, Thermo Scientific
Tubing Transfer Tubing
Nalgene® Pharma-grade Platinum-cured silicone tubing is flexible, durable, and translucent.
  • High-purity tubing
  • Excellent elastic memory
  • Ideal for a variety of transfer applications such as pharmaceutical, laboratory, and bioprocess manufacturing
Resiliency coupled with low hysteresis and compression set ensure that fluid transfers are reliable and accurate every time.
To fit in a wide range of applications, the new Nalgene® Pharma-Grade Platinum-cured silicone tubing can be sterilized by most methods, including autoclaving, gamma-irradiation, and EtO.
Certifications: The following tests were performed on the final extruded product: USP 87, USP 88, ISO10993-3, and ISO10933-4.
Tubing is manufactured, double-bagged, and sealed in an ISO Class 7 clean room. It meets EP 3.1.9 silicone elastomer for closures and tubing and USP 661 Physlochemical tests. A validation binder is available with test reports on the final extruded product that was irradiated and autoclaved to mimic real-use conditions.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty