Would you like to log in again?
Parasite Suspension, Microbiologics
Supplier: Microbiologics
FP02 FP03 FP06 FP08 FP09 FP05 FP04 FP10 FP01 FP11 FP12
89508-180EA 105.97 USD InStock
89508-180 89508-192 89508-182 89508-194 89508-184 89508-174 89508-196 89508-176 89508-188 89508-178 89508-190
Parasite Suspension, Microbiologics
Specimens Live Specimens Animals - Living Material
Parasite Suspensions help labs QC their parasitology diagnostic kits and methods, microscopic examinations, staining procedures, and proficiency programs. Designed to mimic a patient sample for simple control processing, these economical and easy-to-use Parasite Suspensions outperform similar products on the market. They are packaged in a vial containing a 1 ml suspension of a specific parasitic population with known characteristics, formalin and fecal matter. Parasite Suspensions are CE Marked as an In-Vitro Diagnostic (IVD) Medical Device and include a Certificate of Performance which provides detailed information about the parasite.
  • Convenient test ready format saves you time and money.
  • Designed to mimic a patient sample for simple control processing.
  • Includes a Certificate of Performance that provides a detailed description of the specimen.
  • CE Marked as an In-Vitro Diagnostic (IVD) Medical Device.
  • 2 year shelf life.
  • Room temperature storage.
Ordering information: To receive this product, customers must have a valid ATCC End User Licensing Agreement in place with Microbiologics. Registration for this agreement is only required once, per location during the first order. Instructions for how to register can be downloaded from the product page. Go to the "Related Information" box, click on "Additional Product Details", then "Instructions for End User Registration with Microbiologics".
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. Microbiologics Inc. is licensed to use these trademarks and to sell products derived from ATCC® cultures.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty