Would you like to log in again?
Microbiology Quality Control Slides, Microbiologics®
Supplier: Microbiologics
SL01-10
89508-206EA 149.76 USD InStock
89508-206
Microbiology Quality Control Slides, Microbiologics®
Slides Microscopy Control Slides
Microbiology QC Slides come with one or two droplets of an air-dried and methanol-fixed organism, or a single smear containing a designated organism population.
<italic>S
Slides come with a Certificate of Quality and a guaranteed shelf-life of at least three months.
aureus</italic> and <italic>E. coli</italic> Control Slide Microbiology Slides:
Ordering information: To receive this product, customers must have a valid ATCC End User Licensing Agreement in place with Microbiologics. Registration for this agreement is only required once, per location during the first order. Instructions for how to register can be downloaded from our product page. Go to the "Related Information" box, click on "Additional Product Details", then "Instructions for End User Registration with Microbiologics".
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. Microbiologics Inc. is licensed to use these trademarks and to sell products derived from ATCC® cultures.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty