Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VWR® Vacuum Ovens

Supplier: VWR
6292 EWOVWR3-334 EWOVWR215 EWOVWR2-333 EWOVWR316 EWOVWR2-332 6290 6291 EWOVWR214
89508-428EA 13601.15 USD InStock
89508-428 10145-426 10144-916 10145-424 10144-918 10144-910 89508-424 10145-428 89508-426
VWR® Vacuum Ovens
Ovens Vacuum Ovens
The 12.5 L unit features a top-mounted independent evacuation and venting vacuum fittings.

  • Heating system optimizes chamber space for drying, curing, vacuum embedding, and plating applications
  • Polycarbonate safety shield protects door glass
  • Built-in over-temperature protection
  • Vacuum level is displayed on gauges from 0 to 30 in Hg
  • Corrosion-resistant stainless-steel tubing for long-term performance
  • With two removable stacking aluminum shelves
  • Easy-to-clean Type 304 stainless-steel interior chamber
  • Powder-coated heavy-gauge steel exterior for durability
  • Two-year warranty with extended warranty packages available

The 19.8 L and 65.1 L units feature a front-mounted three-way valve for evacuation, venting and purging of inert gases (e.g., Nitrogen or Argon) with vacuum fittings on the front of the unit.

7.6 cm (3") glass wool insulation prevents heat loss. Silicone door gasket and positive latch door maintain seal at all vacuum levels.

Ordering information: Comes with Two-Year Parts and Labor Warranty.

Order Now
 

ORDER

SERVICES


Services: VWRCATALYST™ offers validation services including IQ, OQ, and PQ protocol development and execution, as well as asset management and calibration services (US only). Call 888-793-2300 for more details. Extended warranty plan extends the equipment's original warranty to provide better coverage for your equipment. Each extended warranty is ONLY valid at the point of sale. Extended warranty applies for US only. Each extended warranty includes unlimited labor, parts, and travel as well as priority phone and e-mail support. Service performed by experienced qualified technicians.

White Glove Installation and Verification Service–the white glove installation includes unpacking, inspection, removal/disposal of packing materials, transporting order via elevator and merchandise set up in the room of your choice, installation of casters or leveling feet, power and electrical connections inspected, supply voltage verified, functional and performance verification documented. Our engineers will move the unit from the customer's receiving dock to the lab within the same building. This service does NOT include transportation from vendor warehouse to customer's receiving dock.

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty