Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Arium® Pro Type 1 Water Systems, Sartorius

Supplier: Sartorius

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

arium® pro
H2OPRO-UV-TTOC H2OPRO-VF-BTOC H2OPRO-UV-T-TOC H2OPRO-VF-B-TOC H2OPRO-DI-B H2OPRO-UF-B H2OPRO-UV-BTOC H2OPRO-DI-B H2OPRO-UF-T H2OPRO-VF-B H2OPRO-UV-B H2OPRO-VF-TTOC H2OPRO-DI-T H2OPRO-VF-B H2OBASIC-B H2OPRO-DI-T H2OBASIC-T H2OPRO-VF-T H2OPRO-UF-T H2OPRO-UV-B-TOC H2OPRO-UV-T H2OPRO-UV-T H2OPRO-VF-T H2OPRO-UV-B H2OPRO-VF-T-TOC H2OPRO-UF-B H2OBASIC-T H2OBASIC-B
89510-564EA 7767.95 USD InStock
CA89510-576 CA89510-578 CA89510-556 CA89510-572 CA89510-594 89510-564 89510-588 89510-566 CA89510-590 89510-560 89510-582 CA89510-570 89510-584 CA89510-588 89510-578 CA89510-566 89510-556 89510-558 CA89510-584 CA89510-564 89510-576 89510-570 CA89510-560 CA89510-582 89510-572 89510-594 89510-590 CA89510-558
Arium® Pro Type 1 Water Systems, Sartorius
Water Purification Systems Pure and Ultra Pure Water Systems
The arium pro series consists of flexible systems with a device configuration that can be custom-tailored to your applications and features an outstanding cost-benefit ratio.

  • Arium Delivers 120 L of Ultrapure Water
  • Conductivity: 0.055 Micros/cm
  • Real-Time Monitoring and Data Display
  • Touch-Activated Functions
  • Flexible Device Configuration

Per hour, an arium pro can deliver up to 120 L of high-quality ultrapure water with a conductivity of 0.055 micros/cm (approximately 18.2 MΩ/cm at 25°C).

The integrated real-time monitoring and data display indicates feed conductivity, total water consumption, and the pressure regulator. The control panel features touch-activated functions and intuitive menu navigation.

Certifications: UL/CSA listed.

Accessories information: Accessories are sold separately, and installation is required; please contact your local Avantor representative for more information.

Ordering information: This system includes a two-year manufacturer's warranty.

Order Now
 

ORDER

SPECIFICATIONS

ACCESSORIES

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty