Would you like to log in again?
Arium® Comfort I, Type 1 and 3 Combined Water Systems, Sartorius
Supplier: Sartorius
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
arium® comfort
H2O-I-2-UV-B H2O-I-1-UV-T H2O-I-1-UV-B H2O-I-2-TOC-T H2O-I-1-TOC-B H2O-I-2-UV-T H2O-I-1-UV-B H2O-I-1-T H2O-I-1-UV-T H2O-I-2-B H2O-I-2-B H2O-I-2-TOC-T H2O-I-1-TOC-B H2O-I-1-TOC-T H2O-I-2-UV-T H2O-I-1-T H2O-I-2-TOC-B H2O-I-2-T H2O-I-1-B H2O-I-1-TOC-T H2O-I-1-B H2O-I-2-UV-B H2O-I-2-TOC-B H2O-I-2-T
89510-644EA 10528.66 USD InStock
89510-644 CA89510-632 89510-622 89510-634 CA89510-620 89510-646 CA89510-634 CA89510-622 CA89510-644 89510-640 89510-652 CA89510-652 89510-620 89510-632 CA89510-640 CA89510-628 89510-626 89510-648 89510-638 CA89510-646 89510-628 CA89510-638 CA89510-626 CA89510-648
Arium® Comfort I, Type 1 and 3 Combined Water Systems, Sartorius
Water Purification Systems Pure and Ultra Pure Water Systems
Arium comfort systems produce ASTM Type 1 ultrapure water and Type 3 pure water.
  • Efficient iJust Technology
  • Unique Filter Cartridge
  • Touch-Screen Control Panel
  • UV Lamp Prevents Microbiological Growth
  • Systems Produce ASTM Type 1 and 3 Water
The systems feature the latest reverse osmosis technology combined with electrodeionization (EDI) and a unique filter cartridge for producing ultrapure water of the highest quality, delivering rates up to 120 liters per hour
The systems also feature efficient iJust technology, flexible and space-saving twin technology, and a touch-screen control panel.
The UV lamp (185/254 nm) prevents microbiological growth and reduces the TOC content to a minimum.
Certifications: UL/CSA listed.
Accessories information: Accessories are sold separately, and installation is required; please contact your local Avantor representative for more information.
Ordering information: This system includes a two-year manufacturer's warranty.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty