Would you like to log in again?
Microscope Slide Storage Boxes for 25 Slides, Globe Scientific
Supplier: Globe Scientific
513075B 513075W 513075AST 513075A 513075G 513075N
89510-798EA 5.12 USD InStock
89510-798 89510-800 89510-796 10024-026 89510-802 89510-804
Microscope Slide Storage Boxes for 25 Slides, Globe Scientific
Slide Mailers
Globe's microscope slide storage boxes are designed for the safe and long term storage of microscope slides
An inventory sheet is located on the inside cover for cenvenient slide identification and organization. The base of the box is cork lined for added protection.
They are produced from durable ABS plastic and offer a hinged lid for easy opening and closing.
Ordering information: Available in 4 sizes, these storage boxes accommodate 25 x 75 mm and 3 x 1" microscope slides.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty