Would you like to log in again?
Amplitube™ PCR Reaction Strips, Simport Scientific
Supplier: Simport
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Amplitube™
T320-1G T320-1R T321-2G T321-3N T321-1N T321-1R T321-2B T320-1R T321-1Y T321-2N T321-1R T321-1B T320-1B T320-1N T321-2R T321-2R T321-2N T320-10B T320-10G T321-2G T320-1Y T321-2Y T321-1N T320-10Y T321-1Y T321-1B T320-10R T321-1G T320-1Y T321-2B T320-1G T320-1N T320-10N T321-1G T320-1B T321-1B T321-2Y
60872-640CS 166.18 USD InStock
CA60872-650 60872-640 490010-418 CA60872-644 89511-398 CA60872-641 89429-002 60872-732 89429-000 89511-392 CA48648-506 CA48648-505 CA48648-504 CA48648-503 CA60872-648 89511-396 CA48648-502 89511-394 CA11028-432 CA11028-430 60872-650 89429-006 89429-004 89429-008 89511-400 60872-648 89429-012 60872-642 89429-010 89511-402 60872-644 CA11028-426 89428-998 CA11028-424 CA11028-428 89428-996 CA60872-639
Amplitube™ PCR Reaction Strips, Simport Scientific
Tubes PCR Tubes
PCR Reaction strips include 8 or 12 integral 0.2 ml tubes with ultra-thin sidewalls and bottoms to ensure uniform and efficient temperature transfer.
  • Autoclavable
Non-attached cap strips are available in a dome or flat top design to ensure a perfect closure during the whole thermal cycle. Cap strips are not included and must be ordered separately.
All strips are molded of polypropylene under the most stringent conditions and are offered colorless, or in four different color varieties.
They are more easily handled than single tubes, and will precisely fit standard well spacing. They can also be used with 8- and 12-channel hand-held pipettors.
Certifications: Certified RNase-, DNase-, Pyrogen-, and DNA-free.
Packaging: Packed in tamper-proof resealable bags.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty