Would you like to log in again?
Biohazard Disposal Bags
Supplier: Heathrow Scientific
HS10323 HS10322 HS10321 HS10320
89511-762CS 111.46 USD InStock
89511-762 89511-760 89511-766 89511-764
Biohazard Disposal Bags
Bags Waste Disposal Bags
Biohazard disposal bags are ideal for processing biohazardous materials that require autoclaving prior to disposal.
  • Bags are designed for personal or lab use
  • Available in sizes to suit all needs
  • Autoclavable to 121 °C (250 °F)
The polypropylene bags include an indicator patch that darkens when autoclaved. Handy-size bag holder is ideal for collecting tips and tubes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty