Would you like to log in again?
WHEATON® CryoFile® and CryoFile®XL Storage Boxes, DWK Life Sciences
Supplier: DWK Life Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
WHEATON®
W651602-XL W651610-B W651604-XL W651610-R W651610-P W651610-G W651601-XL W651600-XL W651604 W651601 W651600 W651610-Y W651605 W651603 W651602 W651610-W W651603-XL W651605-XL
89092-390CS 187.66 USD InStock
89092-390 89092-380 89092-392 89092-382 89092-394 89092-384 89511-868 89092-396 89092-386 89092-376 89092-398 89092-388 89092-378 89511-872 89511-870 89511-876 89511-866 89511-874
WHEATON® CryoFile® and CryoFile®XL Storage Boxes, DWK Life Sciences
Boxes Cryoboxes
These CryoFile® and CryoFile®XL storage boxes provide high-quality storage in cryogenic temperatures.
  • Partitions numbered from 1 to 81 for easy content identification
  • Numbering system printed on lid and bottom of box
  • Six colors provide easy sample identification
  • Water repellent allows for longer durability
  • For use with vapor phase of liquid nitrogen
Directional holes in bottom allow for drainage and orientation of bottom to top of box.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty