Would you like to log in again?
2-IN-1 SEAL CAP FOR AGILENT 1050 1100 12
Supplier: OPTIMIZE TECHNOLOGIES, INC.
2-In-1 Seal Cap^ Our 2-In-1 Seal Cap Replaces Both The Oem Outlet Gold Seal And Outlet Cap With A Single Component. Use It On Your Purge Valve, On Your Oem Inlet And Outlet Check Valves, Or Anywhere Else On Your 1200/ 1100/ 1050 System Where Gold Seals And Outlet Caps Are Required.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
2-IN-1 SEAL CAP FOR AGILENT 1050 1100 12 89522-330 27-31-03075 Pack of 4 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty