Would you like to log in again?
DEUTERIUM DOUBLELIFE LAMP WATERS 996 299
Supplier: OPTIMIZE TECHNOLOGIES, INC.
Deuterium Double Life Lamp Waters 996 299^ Optimize Offers A Broad Selection Of Lamps For Oem Hplc Detectors. The High Quality Standard Lamps Come With A Guaranteed 1,000 Hour Lifetime.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
DEUTERIUM DOUBLELIFE LAMP WATERS 996 299 89522-598 39-32-02835 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty