Would you like to log in again?
OPTI-MAX CARTRIDGE CHECK VALVE CONVERSIO
Supplier: OPTIMIZE TECHNOLOGIES, INC.
Opti-Max Cartridge Check Valve Conversion^ Make The Switch To Opti-Max For Less Than Ever Before With Opti-Max Conversion Kits. Each Kit Includes All The Opti-Max Check Valve Housings And Cartridges You Need To Convert Your Hplc Instrument.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
OPTI-MAX CARTRIDGE CHECK VALVE CONVERSIO 89522-610 39-57-02166 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty