Would you like to log in again?
ACTERO™ MediaBox, Foodchek
Supplier: FOODCHEK SYSTEMS INC.
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
FCM-120 FCM-114 FCM-123 FCM-207 FCM-046 FCM-044 FCM-128 FCM-127 FCM-206 FCM-120 FCM-116 FCM-044 FCM-042 FCM-034 FCM-048 FCM-047 FCM-035 FCM-036 FCM-033 FCM-040 FCM-113 FCM-041 FCM-117 FCM-036 FCM-032 FCM-114 FCM-043 FCM-117 FCM-043 FCM-237 FCM-116 FCM-119 FCM-118 FCM-041 FCM-040 FCM-123 FCM-032 FCM-112 FCM-035 FCM-045 FCM-048 FCM-128 FCM-031 FCM-042 FCM-112 FCM-115 FCM-045 FCM-129 FCM-110 FCM-205 FCM-031 FCM-046 FCM-129 FCM-110 FCM-115 FCM-240 FCM-111 FCM-047 FCM-236 FCM-113 FCM-127 FCM-034 FCM-033 FCM-119 FCM-111 FCM-118
89522-688EA 246.88 USD InStock
CA10146-836 CA10146-838 CA10127-234 CA10146-834 CA10127-232 CA10159-100 89522-688 CA89522-698 CA10146-840 CA10146-842 CA89522-696 CA10159-102 10159-098 89522-684 89522-686 89522-680 10159-094 10159-096 10500-814 89522-682 89522-706 10127-232 89522-708 CA10159-104 10127-234 89522-702 CA10159-106 CA89522-678 89522-704 CA89522-712 10159-102 89522-710 CA89522-688 89522-712 10159-104 CA89522-686 89522-678 CA89522-684 10159-100 CA89522-682 CA89522-680 CA89522-708 CA89522-706 CA89522-704 CA89522-702 10146-846 10159-106 CA89522-700 10146-848 CA89522-710 89522-700 10146-850 CA10146-848 10146-838 CA10146-844 10146-834 CA10146-846 10146-836 10146-842 CA10146-850 89522-698 10146-844 10146-840 10500-808 89522-696 10500-806
ACTERO™ MediaBox, Foodchek
Media for Microbiology Prepared Media for Microbiology
The Actero™ MediaBox™ is the solution for laboratories that require liquid culture media on-demand
While Actero Universal Enrichment media represent fast-acting and established formulations that can be used for standard testing protocols employed in today’s labs, the Patent Pending Actero Elite Enrichment Media represent enhanced formulations that further reduce the required enrichment time to an industry-leading low.
Get even faster time-to-results and accuracy when you combine Actero Enrichment Media with the FoodChek™ MICT Reader and Assay
Delivering fresh, sterile media in an easy-to-handle, stackable, storage box, Actero™ MediaBox™ provides affordable, ready-to-use liquid culture media in 5 L and 10 L sizes, with a 20 L option available upon request.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty