Would you like to log in again?
VWR® Sterile Coveralls, 10⁻⁶ SAL
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
VWR®
89523-472 89523-460 89523-466 89523-462 89523-468 10853-872 89523-458 89523-470 89523-464
89523-464CS 427.28 USD InStock
89523-464 89523-462 89523-458 89523-468 89523-466 89523-472 89523-460 10853-872 89523-470
VWR® Sterile Coveralls, 10⁻⁶ SAL
Disposable Garments
VWR® new sterile coveralls provide a solid option for customers requiring a functional garment that is 10-6 SAL
  • Gamma irradiated to SAL 10-6
  • Durable fabric
  • Excellent breathability
  • Suitable for ISO Class 5 or higher cleanrooms
  • Individually bagged
These comfortable coveralls are manufactured from a durable SMS fabric and feature bound seams. Rigorously tested and manufactured in an ISO Certified facility under stringent process controls to ensure that each product meets exacting quality standards and performs to specification. Coveralls are Clean Level II certified according IEST test results, and offer a BFE of over 98%. Designed for performance in a variety of clean environments in multiple markets including Compounding Pharmacies and Biopharmaceutical production.
Certifications: Manufactured in an ISO Class 8 (FED-STD-209E Class 100,000/M3.5) cleanroom.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty