Would you like to log in again?
VWR® C-Leg Bench Frame with Top, Double Bay Uprights and 4 Shelves
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
14-C100VWR474 14-C103VWR439 14-C103VWR472 14-C100VWR438 14-C101VWR437 14-C101VWR438 14-C101VWR474 14-C103VWR436 14-C101VWR439 14-C101VWR473 14-C103VWR475 14-C101VWR475 14-C100VWR472 14-C101VWR436 14-C103VWR437 14-C100VWR437 14-C103VWR474 14-C100VWR439 14-C100VWR473 14-C103VWR438 14-C101VWR472 14-C103VWR473 14-C100VWR436 14-C100VWR475
89525-040EA 7220.23 USD InStock
89525-040 89524-404 89525-038 89524-964 89524-326 89524-898 89525-036 89524-900 89524-328 89524-966 89525-034 89524-968 89524-902 89524-264 89524-330 89524-398 89524-332 89524-266 89524-400 89524-402 89524-896 89524-962 89524-260 89524-262
VWR® C-Leg Bench Frame with Top, Double Bay Uprights and 4 Shelves
Furniture Tables and Workbenches
VWR® lab benches are ergonomically designed to meet the needs of laboratory personnel.
  • Ergonomic and highly adaptable to the needs of the user
  • Flexible, modular design
  • Suitable for heavy loads up to 1100 lbs.
  • Offering a wide range of accessories to meet any need
There are many possible adjustments, all components fit together seamlessly, and each detail is designed for real world functionality. The lab bench adapts to the work process and the needs of the user. As a result, all movements of the user can be optimized, which in turn increases performance and productivity and decreases work-related stress. VWR® lab benches are height adjustable in manual, motorized and hand crank versions. All bench frames can be equipped with casters to create a mobile workstation solution.
Ordering information: Configurations include: frame, top, double bay uprights, 4 x 12"D Shelves. Frame color is light neutral. Top includes black phenolic or black laminate options.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty