Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VWR® C-Leg Bench Frame with Top

Supplier: VWR

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

14-C100VWR122 14-C100VWR123 14-C103VWR444 14-C101VWR125 14-C103VWR125 14-C101VWR121 14-C101VWR403 14-C100VWR442 14-C103VWR442 14-C100VWR444 14-C103VWR121 14-C103VWR124 14-C103VWR403 14-C103VWR123 14-C103VWR441 14-C101VWR440 14-C101VWR441 14-C100VWR441 14-C100VWR121 14-C101VWR123 14-C101VWR122 14-C100VWR443 14-C101VWR124 14-C101VWR442 14-C100VWR403 14-C100VWR125 14-C103VWR122 14-C101VWR444 14-C100VWR445 14-C101VWR445 14-C103VWR440 14-C103VWR445 14-C100VWR124 14-C103VWR443 14-C101VWR443 14-C100VWR440
89524-906EA 4844.95 USD InStock
89524-906 89524-908 89524-206 89524-826 89524-828 89524-904 89524-198 89524-992 89524-970 89524-972 89524-334 89524-356 89524-336 89524-974 89524-830 89524-192 89524-194 89524-270 89524-196 89524-272 89524-976 89524-338 89524-832 89524-978 89524-834 89524-836 89524-838 89524-220 89524-340 89524-200 89524-342 89524-202 89524-268 89524-840 89524-842 89524-204
VWR® C-Leg Bench Frame with Top
Furniture Tables and Workbenches
VWR® lab benches are ergonomically designed to meet the needs of laboratory personnel.

  • Ergonomic and highly adaptable to the needs of the user
  • Flexible, modular design
  • Suitable for heavy loads up to 1100 lbs.
  • Offering a wide range of accessories to meet any need
  • 120V, 60Hz Electrical Specifications (Motorized Models)

There are many possible adjustments, all components fit together seamlessly, and each detail is designed for real world functionality. The lab bench adapts to the work process and the needs of the user. As a result, all movements of the user can be optimized, which in turn increases performance and productivity and decreases work-related stress. VWR® lab benches are height adjustable in manual, motorized and hand crank versions. All bench frames can be equipped with casters to create a mobile workstation solution.

Ordering information: Configurations include frame and top. Bench color is light neutral. Top includes black phenolic or black laminate options.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty