Would you like to log in again?
Fluid D, BD Life Sciences
Supplier: BD Life Sciences
210049 210049
90000-000PK 799.35 USD InStock
CA90000-000 90000-000
Fluid D, BD Life Sciences
Media for Microbiology Prepared Media for Microbiology
Fluid D is used for diluting or rinsing samples containing lecithin or oil when performing sterility testing.
  • Peptone water helps dilute and rinse
  • Inclusion of polysorbate 80 acts as a surfactant
  • Nontoxic
Fluid D is peptone water with polysorbate 80, and is used for diluting or rinsing when performing sterility testing. This fluid aids in the complete rinsing of the membrane filter apparatus and is not toxic to microorganisms. Fluid D contains polysorbate 80, which acts as a surfactant to break down the lecithin or oils present.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty