Would you like to log in again?
Difco™ Tetrathionate Broth Base, BD Biosciences
Supplier: BD Biosciences
210420 210430 210430 210420
90000-006EA 551.24 USD InStock
90000-006 CA90000-008 90000-008 CA90000-006
Difco™ Tetrathionate Broth Base, BD Biosciences
Media for Microbiology Additives and Supplements for Microbiology
Tetrathionate Broth Base is used as a selective enrichment medium for the isolation of Salmonella from feces, urine, foods, and other materials of sanitary importance.
  • Peptones provide nitrogen, vitamins, amino acids, and carbon
  • Oxgall inhibits Gram-positive microorganisms
  • Calcium carbonate neutralizes and absorbs toxic metabolites
Tetrathionate, which is formed in the medium by the addition of the iodine-iodide solution, inhibits the normal intestinal flora of fecal specimens.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty