Would you like to log in again?
BBL™ Blood Agar Base (Infusion Agar), BD
Supplier: BD Biosciences
211037 211037
90000-048EA 417.88 USD InStock
CA90000-048 90000-048
BBL™ Blood Agar Base (Infusion Agar), BD
Media for Microbiology Dehydrated Media for Microbiology
Blood Agar Base (Infusion Agar), with the addition of sterile blood, is used for the isolation, cultivation, and detection of hemolytic activity of streptococci and other fastidious microorganisms.
  • All-purpose medium
  • Heart muscle infusion, casein peptone, and yeast extract provide nutrients
  • Sodium chloride maintains osmotic equilibrium
  • Agar is the solidifying agent
Infusion Agar is an all-purpose medium which has been used for many years as a base for the preparation of blood agars. Supplementation with blood (5-10%) provides additional growth factors for fastidious microorganisms, and is the basis for determining hemolytic reactions.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty