Would you like to log in again?
BBL™ Brucella Broth, BD Biosciences
Supplier: BD Biosciences
BBL™
211088
90000-076EA 284.63 USD InStock
90000-076
BBL™ Brucella Broth, BD Biosciences
Media for Microbiology Dehydrated Media for Microbiology
Brucella Broth is used for the cultivation of Brucella species and for the isolation and cultivation of a wide variety of fastidious and nonfastidious microorganisms.
  • Use for the isolation and cultivation of a wide variety of microorganisms
  • Isolates and cultivates nutritionally fastidious specimens
  • Can also be used to cultivate Campylobacter
This medium is recommended for the cultivation of Brucella species and was recommended as one Brucella Media of several media suitable for use as the liquid medium component of biphasic blood culture bottles.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty