Would you like to log in again?
BBL™ CTA Medium™, BD Biosciences
Supplier: BD Biosciences
211096 211096
90000-080EA 352.59 USD InStock
CA90000-080 90000-080
BBL™ CTA Medium™, BD Biosciences
Media for Microbiology Dehydrated Media for Microbiology
CTA Medium™ (Cystine Trypticase™ Agar Medium) is a simple, basic medium for the maintenance and detection of motility of a wide variety of microorganisms.
  • Cystine and casein peptone provide necessary nutrients
  • Carbohydrate fermentation is detected by a visible color change
  • Used for the detection of bacterial motility
With added carbohydrates, it is useful for the determination of fermentation reactions.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty