Would you like to log in again?
BBL™ Cary-Blair Transport Medium, BD
Supplier: BD Biosciences
211102
90000-084EA 281.04 USD InStock
90000-084
BBL™ Cary-Blair Transport Medium, BD
Media for Microbiology Dehydrated Media for Microbiology
Cary-Blair Transport Medium is a semisolid medium recommended for use in the transportation and preservation of clinical specimens, primarily stool and rectal swabs.
  • Sodium thioglycollate is added to impede oxidation
  • Disodium phosphate acts as a buffering agent
  • Alkaline pH minimizes destruction of bacteria due to acid formation
  • Low nutrient content
This medium possesses a low nutrient content, a low oxidation-reduction potential, and a high pH, making it successful in recovering Salmonella and Shigella.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty