Would you like to log in again?
BBL™ Columbia Agar, BD
Supplier: BD Biosciences
215191 211125 211125
95038-622PK 180.85 USD InStock
95038-622 90000-094 CA90000-094
BBL™ Columbia Agar, BD
Media for Microbiology Dehydrated Media for Microbiology
Columbia Agar Base is a highly nutritious, general-purpose medium for the isolation and cultivation of nonfastidious and fastidious microorganisms from a variety of clinical materials.
  • Used with blood in isolating and cultivating fastidious microorganisms
  • Used in qualitative procedures
  • For isolating and cultivating Haemophilus species from clinical specimens
Columbia Agar Base is utilized as the base for media containing blood and for selective media formulations in which various combinations of antimicrobial agents are used as additives.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty