Would you like to log in again?
BBL™ Endo Agar, BD
Supplier: BD Life Sciences
211199 211199
90000-114EA 348.92 USD InStock
CA90000-114 90000-114
BBL™ Endo Agar, BD
Media for Microbiology Prepared Media for Microbiology
Endo Agar is a differential and slightly selective culture medium for the detection of coliform and other enteric microorganisms.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty