Would you like to log in again?
BD BBL™ Eosin Methylene Blue Agar, BD Diagnostics
Supplier: BD Diagnostics
211215 211215 211221 211221
90000-118EA 232.49 USD InStock
CA90000-118 90000-118 CA90000-122 90000-122
BD BBL™ Eosin Methylene Blue Agar, BD Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
This dehydrated culture media is used for isolating and differentiating lactose-fermenting from lactose-nonfermenting gram-negative enteric bacilli.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty