Would you like to log in again?
BBL™ M-Green Yeast and Mold Broth, BD Biosciences
Supplier: BD Biosciences
211287 211286 211287 211286
90000-142EA 432.68 USD InStock
90000-142 CA90000-140 90000-140 CA90000-142
BBL™ M-Green Yeast and Mold Broth, BD Biosciences
Media for Microbiology Dehydrated Media for Microbiology
M-Green Yeast and Mold Broth is used for testing soft drinks and other beverages for growth of yeast and mold.
  • Present yeast produces large green, opaque colonies
  • Mold colonies are green and filamentous
  • Used to test food and beverages
BBL™ M-Green Yeast and Mold Broth is used for selective testing of yeast and mold by pH.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty