Would you like to log in again?
Difco™ Malt Extract Broth, BD Diagnostics
Supplier: BD Diagnostics
Difco™
211320 211320
90000-150EA 194.66 USD InStock
CA90000-150 90000-150
Difco™ Malt Extract Broth, BD Diagnostics
Media for Microbiology Prepared Media for Microbiology
Malt Extract Broth is used for cultivating yeasts and molds.
  • Malt extract provides carbon, protein, and nutrient sources for essential growth
  • Maltose is an energy source
  • Dextrose is included as a source of fermentable carbohydrate
  • Yeast extract provides vitamins and additional nitrogen and carbon
The acidic pH of Malt Extract Broth allows for the optimal growth of molds and yeasts while restricting bacterial growth.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty