Would you like to log in again?
MR-VP (Methyl Red Voges Proskauer) Broth, BD Life Sciences
Supplier: BD Life Sciences
216300 216300 221667 211383 221667 211383
90001-446PK 36.22 USD InStock
90001-446 CA90001-446 90000-924 CA90000-924 CA90000-166 90000-166
MR-VP (Methyl Red Voges Proskauer) Broth, BD Life Sciences
Media for Microbiology Dehydrated Media for Microbiology
MR-VP (Methyl Red Voges Proskauer) Broth is recommended for use in the performance of Voges-Proskauer and Methyl Red tests as an aid in the identification of enteric Gram-negative bacilli.
  • Methyl red is yellow at pH 6.2 and above, and red at pH 4.4 and below
  • Particularly useful in the identification of Enterobacteriaceae
The same broth is used for both tests. The methyl red test detects production of acids formed during metabolism using mixed acid fermentation pathway with pyruvate as a substrate. The VP test uses alpha-naphthol and potassium hydroxide to test for the presence of acetoin.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty