Would you like to log in again?
BBL™ Mannitol Salt Agar, BD
Supplier: BD Biosciences
BBL™
211407 211407 211410 211410
90000-182EA 131.93 USD InStock
90000-182 CA90000-184 CA90000-182 90000-184
BBL™ Mannitol Salt Agar, BD
Media for Microbiology Dehydrated Media for Microbiology
Mannitol Salt Agar is a selective and differential medium for the detection and enumeration of staphylococci from clinical and nonclinical specimens.
  • Beef extract and peptones provide essential growth factors
  • 7.5% concentration of sodium chloride inhibits bacterial growth
  • Mannitol fermentation aids in differentiation of staphylococcal species
Difference in colony growth colors also acts as an identifying feature.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty