Would you like to log in again?
BBL™ Moeller Decarboxylase Broth Base with Amino Acids, BD Life Sciences
Supplier: BD Life Sciences
BBL™
221661 221661 221659 211430 211430
90001-436PK 45.27 USD InStock
90001-436 90001-438 CA90001-438 90000-186 CA90000-186
BBL™ Moeller Decarboxylase Broth Base with Amino Acids, BD Life Sciences
Media for Microbiology Prepared Media for Microbiology
Moeller Decarboxylase Broth Base, when supplemented with arginine, aids in the differentiation of enteric bacilli by means of a dihydrolase reaction.
  • Differentiates gram-negative enteric bacilli based on decarboxylation of amino acids
  • Beef extract and peptic digest of tissue provide nitrogenous nutrients
  • Dextrose is a fermentable carbohydrate
  • Pyridoxal is the co-factor for the decarboxylase enzyme
  • Bromo cresol purple and cresol red are pH indicators
Arginine is first hydrolyzed to ornithine, which is then decarboxylated to form putrescine. Formation of these amines increases the pH of the medium, changing the color of the indicator from yellow to purple. If the organisms do not produce the appropriate enzyme, the medium remains acidic, yellow in color. Each isolate to be tested should also be inoculated into Moeller Decarboxylate Broth Base medium tube lacking the amino acid.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty