Would you like to log in again?
Mueller Hinton Agar II, BD
Supplier: BD Diagnostics
211441 211441 211438 212257 211438
90000-192EA 1051.56 USD InStock
90000-192 CA90000-190 90000-190 CA90000-192 90000-572
Mueller Hinton Agar II, BD
Media for Microbiology Dehydrated Media for Microbiology
Mueller-Hinton agar is a microbiological growth medium that is commonly used in antibiotic susceptibility testing.
  • Non-selective, non-differential medium
  • Contains starch
  • Loose agar for better diffusion of antibiotics
  • Truer zone of inhibition
Agar is also used to isolate and maintain Neisseria and Moraxella species.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty