Would you like to log in again?
Mycosel™ Agar Slants, BD
Supplier: BD Diagnostics
BBL™
221130 211462 220966 220966 220967 211462 220967
90001-168CS 508.14 USD InStock
CA90000-202 90001-168 CA90001-166 90000-202 CA-90001-168 90001-240 90001-166
Mycosel™ Agar Slants, BD
Media for Microbiology Prepared Media for Microbiology
Mycosel™ Agar is a selective medium for the isolation of pathogenic fungi from materials having a mixed flora of other fungi and bacteria.
  • Soybean meal peptones provide nutrients
  • Dextrose is an energy source
  • Cycloheximide inhibits most saprophytic molds
  • Chloramphenicol inhibits most gram-positive and gram-negative bacteria
Mycosel™ Agar was developed by using the ingredients of Mycophil™ Agar as a nutritive base to which cycloheximide and chloramphenicol were added as selective agents. It is widely used for the isolation of fungi from a variety of sources, and is recommended for the recovery of dermatophytes.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty