Would you like to log in again?
BBL™ Phosphate Buffer, pH 7.2, BD
Supplier: BD
211544 211544
90000-224EA 221.37 USD InStock
CA90000-224 90000-224
BBL™ Phosphate Buffer, pH 7.2, BD
Media for Microbiology Dehydrated Media for Microbiology
Phosphate Buffer, pH 7.2 is used for the preparation of dilution blanks for use in the examination of waters, dairy products, foods, and other materials.
  • Sodium carbonate is a pH regulator
  • Prepared appearance: Colorless to light yellow and clear to trace hazy
Phosphate buffer is used in the preparation of dilution blanks for use in microbiological testing rather than unbuffered water in order to standardize this potential variable due to the wide variation in the pH of puriifed water from multiple sources.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty