Would you like to log in again?
BBL™ Spirolate Broth, BD Biosciences
Supplier: BD Biosciences
211636
90000-248EA 168 USD InStock
90000-248
BBL™ Spirolate Broth, BD Biosciences
Media for Microbiology Dehydrated Media for Microbiology
BBL™ Spirolate Broth was developed for mass culture of the Reiter treponeme in a medium without agar.
  • Various extracts supply nitrogenous growth factors
  • Contains everything required for the metabolism of Reiter treponemes
  • Sodium chloride aids in the maintenance of the osmotic equilibrium of the medium
  • Supplementation with fatty acids
  • Stimulatory effect on growth of the Reiter treponeme
Spirolate Broth can also be used for cultivating other spirochetes.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty