Would you like to log in again?
Difco™ Sabouraud Dextrose Agar, BD
Supplier: BD Diagnostics
Difco™
211661 211661 210950 210950 210930
90000-258EA 670.61 USD InStock
CA90000-258 90000-258 90000-038 CA90000-039 90000-020
Difco™ Sabouraud Dextrose Agar, BD
Media for Microbiology Dehydrated Media for Microbiology
Sabouraud Dextrose Agar is used in qualitative procedures for the cultivation of dermatophytes and other pathogenic and nonpathogenic fungi from clinical and nonclinical specimens.
  • Low pH is favorable for the growth of fungi
  • Dehydrated appearance: Light beige, free-flowing
  • Peptones are the source of nitrogenous growth factors
The addition of antimicrobics further increases the selectivity of the medium.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty