Would you like to log in again?
Difco™ Nutrient Agar, BD
Supplier: BD Diagnostics
211665 213000 211665 212000 212000 213000
90000-744EA 328.11 USD InStock
90000-744 CA90000-476 CA90000-744 CA90000-260 90000-260 90000-476
Difco™ Nutrient Agar, BD
Media for Microbiology Dehydrated Media for Microbiology
Nutrient Agar is used for the cultivation of bacteria and for the enumeration of organisms in water, sewage, feces, and other materials.
  • Beef extract provides carbohydrates, vitamins, nitrogen, and salts
  • Peptones provide a source of nitrogen
  • Agar is the solidifying agent
Nutrient Agar consists of peptone, beef extract, and agar. This relatively simple formulation provides the nutrients necessary for the replication of a large number of microorganisms that are not excessively fastidious.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty