Would you like to log in again?
BBL™ TSN (Trypticase Sulfite Neomycin) Agar, BD
Supplier: BD Biosciences
211690
90000-276EA 490.73 USD InStock
90000-276
BBL™ TSN (Trypticase Sulfite Neomycin) Agar, BD
Media for Microbiology Dehydrated Media for Microbiology
TSN Agar is used for the selective isolation of clostridium perfringens.
  • Effectively isolates Clostridium perfringens
  • Fine
  • Homogenous
Neomycin and polymyxin are inhibitory for Gram-negative enteric bacilli, and neomycin at the concentration employed at least partially inhibits C. bifermentans.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty