Would you like to log in again?
BBL™ Thioglycollate Medium, Brewer Modified, BD Life Sciences
Supplier: BD Diagnostics
211716 211716
90000-294EA 205.21 USD InStock
CA90000-294 90000-294
BBL™ Thioglycollate Medium, Brewer Modified, BD Life Sciences
Media for Microbiology Additives and Supplements for Microbiology
Thioglycollate Medium, Brewer Modified is used for testing sterility of biological products and for the isolation of aerobic and anaerobic organisms.
  • Modified for sterility testing of products containing mercury preservatives
  • Peptones, beef extract, and yeast extract provide essential nutrients
  • Suitable for luxuriant growth of fastidious microorganisms
  • Very small amount of agar helps in maintaining anaerobic condition
The modified medium contains more thioglycollate and is recommended for sterility testing procedure.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty