Would you like to log in again?
Difco™ Tomato Juice Agar, BD Diagnostics
Supplier: BD Biosciences
211794 211794
90000-358EA 916.24 USD InStock
90000-358 CA90000-358
Difco™ Tomato Juice Agar, BD Diagnostics
Media for Microbiology Additives and Supplements for Microbiology
Tomato Juice Agar is used for cultivating and enumerating Lactobacillus species.
  • Tomato juice is a source of carbon, protein, and nutrients
  • Peptone provides a source of nitrogen, amino acids, and carbon
  • Peptonized milk contains lactose as an energy source
  • Agar is the solidifying agent
Tomato Juice Agar is especially useful in cultivating L
Tomato Juice Agar, Special is recommended for the direct plate count of lactobacilli from saliva and for cultivation of other acidophilic microorganisms. This medium is more selective for lactobacilli than Tomato Juice Agar.
acidophilus from clinical specimens and foodstuffs.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty