Would you like to log in again?
Tryptic Soy Broth (TSB) Dehydrated and Prepared Culture Media, BD Diagnostics
Supplier: BD Diagnostics
211772 299501 292770 214886 221823 211768 221716 221092 221351 221092 221351 221715 211768 221093 221823 211823 221815 299936 257203 211822 211822 211823 257160 221815 214889 211771 221715 211825 214887 257202 211772 221716 211771 211824 211825
89125-602PK 358.35 USD InStock
89125-602 CA90000-346 90000-378 CA90000-344 CA-90001-496 CA90000-348 89049-060 CA90001-494 CA90001-224 90001-534 90006-098 CA-90001-332 CA90000-374 89125-838 90000-344 CA90000-372 CA90000-378 90000-348 CA90000-376 90000-346 89125-597 90001-494 90001-496 90001-332 90001-542 90001-224 90001-226 CA90001-534 95038-634 CA-90001-542 90000-372 90004-410 95052-892 90006-602 90000-374
Tryptic Soy Broth (TSB) Dehydrated and Prepared Culture Media, BD Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
Tryptic soy broth is used in microbiology laboratories as a culture broth to grow aerobic bacteria.
  • Used in a wide variety of procedures
  • May be used for suspension and enrichment
  • For cultivation of strains isolated on other media
With the addition of 6.5% sodium chloride, differentiation between salt-tolerant and salt-intolerant species is achieved.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty