Would you like to log in again?
BBL™ Hemoglobin Solution 2%, BD
Supplier: BD Biosciences
211874 211874
90000-410PK 569.28 USD InStock
CA90000-410 90000-410
BBL™ Hemoglobin Solution 2%, BD
Media for Microbiology Dehydrated Media for Microbiology
Hemoglobin solution is ready to use to enrich a variety of mediums for the cultivation of fastidious organisms.
  • Contains an essential growth factor for some Haemophilus species
  • More selective formulations of chocolate agar can be prepared by adding antimicrobial solutions
  • To inhibit the growth of unwanted organisms
This hemoglobin solution is prepared as dehydrated culture media.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty