Would you like to log in again?
BBL™ Bovine Albumin, BD
Supplier: BD Biosciences
211968 211968
90000-474PK 421.6 USD InStock
CA90000-474 90000-474
BBL™ Bovine Albumin, BD
Media for Microbiology Dehydrated Media for Microbiology
Bovine serum albumin, also known as Fraction V, is a serum albumin protein derived from cows that is often used as a protein concentration standard in lab experiments.
  • Numerous possible applications
  • For ELISAs, immunoblots, and immunohistochemistry
  • Can be used as a nutrient in cell and microbial culture
  • Used to stabilize some enzymes during the digestion of Dna
  • To prevent adhesion of the enzyme to reaction vessels
Bovine albumin is used because of its ability to increase signal in assays, its lack of effect in many biochemical reactions, and its low cost.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty