Would you like to log in again?
Difco™ Marine Agar 2216, BD Diagnostics
Supplier: BD Diagnostics
Difco™
212185 212185
90000-518EA 406.18 USD InStock
CA90000-518 90000-518
Difco™ Marine Agar 2216, BD Diagnostics
Media for Microbiology Prepared Media for Microbiology
Marine Agar 2216 Agar is used for cultivating heterotrophic marine bacteria.
  • Peptone and yeast extract provide nitrogen, vitamins, and minerals
  • High salt content helps simulate sea water
  • Agar is the solidifying agent
Numerous minerals are also included to duplicate the major mineral composition of sea water.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty