Would you like to log in again?
BBL™ CLED Agar, BD
Supplier: BD Biosciences
212218
90000-534EA 310.91 USD InStock
90000-534
BBL™ CLED Agar, BD
Media for Microbiology Dehydrated Media for Microbiology
BBL™ CLED Agar is recommended for the isolation, enumeration, and presumptive identification of urinary pathogens on the basis of lactose fermentation.
  • Controls the swarming of Proteus specimens
  • Supports the growth of all potential urinary pathogens and other contaminants
CLED is recommended for use in spread plate technique for detection of bacteria in urine.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty