Would you like to log in again?
BBL™ Mycoplasma Enrichment without Penicillin, BD Biosciences
Supplier: BD
212292
90000-584PK 2030.58 USD InStock
90000-584
BBL™ Mycoplasma Enrichment without Penicillin, BD Biosciences
Media for Microbiology Dehydrated Media for Microbiology
Mycoplasma Enrichment without Penicillin is a lyophilized blend of ingredients used to supplement suitable media when isolation of Mycoplasma is desired.
  • Lyophilized
  • Supplement to Mycoplasma Agar Base and Mycoplasma Broth Base
When used as a supplement, the enrichment enhances the growth of mycoplasmas, and the addition of penicillin inhibits bacteria in mixed cultures.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty