Would you like to log in again?
BBL™ Trypticase™ Soy Agar (TSA) Dehydrated and Prepared Culture Media, BD Biosciences
Supplier: BD Biosciences
BBL™
221087 221082 212263 211043 212305 221086 297941 221283 212305 211046 211047 211764 297941 211047 292762 221803 221082 211046 211764 221185 221803 299623 211043 212263 221288 221287 221086 221283 221288
90000-338EA 237.85 USD InStock
90000-338 CA90001-290 90001-286 90001-220 CA-90005-052 90001-222 90001-218 CA90000-052 CA90000-050 90008-004 CA90001-528 CA90000-592 CA90000-054 CA90000-574 CA90000-338 90001-290 CA-90001-286 90005-242 CA90001-220 CA90001-254 90000-050 90005-052 90000-592 90001-528 90000-054 CA90001-218 CA-90001-288 90000-052 90000-574
BBL™ Trypticase™ Soy Agar (TSA) Dehydrated and Prepared Culture Media, BD Biosciences
Media for Microbiology Dehydrated Media for Microbiology
Trypticase Soy Agar is a general-purpose medium for microbiology which supports the growth of fastidious as well as nonfastidious microorganisms.
  • Contains casein and soy peptones for a source of general growth
  • Sodium chloride maintains osmotic equilibrium
Lecithin and polysorbate 80, two commonly used neutralizers, are reported to inactivate residual disinfectants when the sample is being collected.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty