Would you like to log in again?
BBL™ MacConkey II Agar, BD
Supplier: BD Life Sciences
BBL™
212306 212306 292861
90004-448EA 2982.75 USD InStock
90004-448 CA90000-594 90000-594
BBL™ MacConkey II Agar, BD
Media for Microbiology Prepared Media for Microbiology
MacConkey II Agar is a selective and differential medium for the detection of coliform organisms and enteric pathogens.
  • Low concentration of bile salts
  • Crystal violet inhibits growth of gram-positive bacteria
  • Lactose and neutral red indicator aid in differentiation of enteric microorganisms
MacConkey II Agar is recommended for use with clinical specimens likely to contain mixed microbial flora, such as urine, because it allows a preliminary gropuing of enteric and other gram-negative bacteria.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty